Would you like to log in again?
ColorpHast™ Strips, pH 5 to 10
Supplier: EMD Millipore
9588-3
470203-340EA 132.79 USD InStock
470203-340
ColorpHast™ Strips, pH 5 to 10
Test Paper pH Indicator Papers and Test Strips
These pH-indicator strips are non-bleeding.
  • Non-Bleeding
  • Non-Contaminating
  • Ideal for All Media in Environmental Analysis
  • Also Suited for Industrial Controls
They are suited for all media in environmental analysis and in industrial, in-process controls.
The strips don't contaminate media.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty