Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Whatman™ Pure Quartz Air Sampling Filter, Grade QM-H, Whatman products (Cytiva)

Supplier: Whatman products (Cytiva)

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Whatman™
1853-150-50 1853-150-50 1853-050-50 1853-047-50 1853-047-50 1853-037-50 1853-090-50 1853-090-50 1853-050-50 1853-037-50
10828-404PK 250.08 USD InStock
10828-404 CA10828-402 CA10828-400 10828-402 CA10827-974 CA10828-404 10827-974 10828-400 CA10828-398 10828-398
Whatman™ Pure Quartz Air Sampling Filter, Grade QM-H, Whatman products (Cytiva)
Filters Glass Fiber Filters
Whatman™ QM-H is a pure quartz fibre filter for use in testing acidic gases, stacks, flues and aerosols. It has low heavy metal content so it is well suited for heavy metals analysis.

  • High stability up to 900 °C
  • High filtration efficiency
  • Binder-free
  • Minimal reactivity with biological or chemical components

Pure quartz fiber filters come in 50-piece packs with five diameters: 37 mm, 47 mm, 50 mm, 90 mm, 150 mm.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty