Would you like to log in again?
5-Hydroxymethylcytosine DNA ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
900-225-0001
75786-620EA 675.09 USD InStock
75786-620
5-Hydroxymethylcytosine DNA ELISA Kit, Enzo Life Sciences
Assays ELISAs
Highly sensitive, rapid ELISA kit to quantify 5-hydroxymethylcytosine in DNA with results that closely correlate to mass spectrometry analysis.
  • Rapid: Fully quantitative results in less than three hours
  • Convenient: User-friendly alternative to mass spectrometry
  • Simple: Efficient and well-established protocol
  • High-throughput: 96-well format ideal for global detection, tissue-specific quantitation, high-throughput compound screening, and more
For the quantitative determination of species independent 5-hydroxymethylcytosine DNA in a variety of DNA samples.
The 5-Hydroxymethylcytosine DNA ELISA kit is a complete, colorimetric, immunometric immunoassay kit for the quantitative determination of 5-hydroxymethylcytosine in DNA  with results in just three hours.
Ordering information: Contains Microtiter Plate, Coating Buffer, ELISA Buffer, Primary Antibody, Antibody-conjugate,  HRP Developer, Controls.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty