Would you like to log in again?
KIM-1 (Human) ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
900-226-0001
75786-622EA 593.39 USD InStock
75786-622
KIM-1 (Human) ELISA Kit, Enzo Life Sciences
Assays ELISAs
Ultra sensitive (1.2 pg/ml) ELISA kit, enabling reduced input sample and matrix interference, for the quantification of KIM-1, an early biomarker for kidney injury or disease.
  • Sensitive: Reliably measure as little as 1.2 pg/ml of KIM-1
  • High Throughput: Results in less than two hours from up to 38 samples in duplicate
  • Specific: Low cross-reactivity to similar proteins
  • Quantitative: Fully quantitative results for both normal and disease state samples that surpass semi-quantitative Western blot analysis
For the quantitative determination of human KIM-1 in urine.
The KIM-1 (human) ELISA kit is a complete, colorimetric, immunometric immunoassay kit for the quantitative determination of human KIM-1 in urine with results in less than two hours.
Ordering information: Contains Microtiter Plate, Assay Buffer, Standard, Antibody-conjugate, Streptavidin-HRP, TMB Substrate, Stop Solution, Wash Buffer Concentrate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty