Would you like to log in again?
ExoPure™ 0.4 µm Immunobeads, Overall Exosome Isolation, Cell Media, BioVision Inc
Supplier: BioVision
AB286904-10TEST
75799-528EA 2275.11 USD InStock
75799-528
ExoPure™ 0.4 µm Immunobeads, Overall Exosome Isolation, Cell Media, BioVision Inc
Assays Cellular Assays
0.4 micron Immunobeads to capture the overall exosome population from cell culture media.
  • 0.4 micron Immunobeads for overall exosome isolation from cell culture media
  • Small sample volume of cell culture medium
  • No ultracentrifugation or other methods for exosome purification required
  • Ready to use and stable for long term (up to 8 months)
  • Suitable for nucleic acid extraction, protein profiling of exosome markers, exosome elution from immunobeads
  • A mouse monoclonal antibody has been used for coating
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty