Would you like to log in again?
NGAL (Monkey) ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BPD-KIT-045
75786-322EA 2039.18 USD InStock
75786-322
NGAL (Monkey) ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of NGAL in monkey urine, plasma, serum, tissue extracts, or culture supernatants.
  • Ready-to-use: Pre-coated plate and individual calibrators save time and reduce errors
  • Easy-to-use: Convenient storage at 4 °C with room temperature incubations
  • Rapid: Results in just four hours from up to 40 samples in duplicate
A colorimetric immunometric ELISA kit for the rapid measurement of NGAL in pig samples. Absorbance is read at 450 nm.
Ordering information: Contains Microtiter Plate, Sample Diluent Conc., Monkey NGAL Calibrator Stock, Wash Solution Conc., Biotinylated Monkey-NGAL Antibody, HRP-Streptavidin, TMB Substrate, Stop Solution. Manufactured by BioPorto Diagnostics.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty