Would you like to log in again?
CRP (Human) ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT102-0001
75786-324EA 429.24 USD InStock
75786-324
CRP (Human) ELISA Kit, Enzo Life Sciences
Assays ELISAs
Sensitive (12.685ng/ml) ELISA kit for the quantification of CRP, an early biomarker for inflammation.
  • Sensitive measurement of CRP, detecting as little as 12.685 ng/ml
  • Negligible reactivity for similar family members
  • High throughput format with results in <2 hours for up to 40 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of human C-reactive protein in serum and plasma.
The CRP (human) ELISA kit is a complete, colorimetric, competitive immunoassay kit for the quantitative determination of human CRP in serum and plasma with results in less than two hours.
Ordering information: Contains Microtiter Plate, Assay Buffer, Standard, Conjugate, Antibody, Streptavidin-HRP, TMB Substrate, Stop Solution, Wash Buffer Concentrate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty