Would you like to log in again?
ExoStd™ Lyophilized Exosome Standard, 100 µg, Human Serum, BioVision Inc
Supplier: BioVision
ExoStd™
AB286846-6VIAL
75799-498EA 4402.89 USD InStock
75799-498
ExoStd™ Lyophilized Exosome Standard, 100 µg, Human Serum, BioVision Inc
Standards Biological Standards
Lyophilized and purified Exosomes available from Human Serum (30 µg).
  • Purified and lyophilized exosomes
  • Control standards for multiple applications including FACS, WB, ELISA
  • Calibration standards for quantitation of exosome-derived markers from biological samples
  • Easy to ship and store
  • Stable for over 36 months at 4°C
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty