Would you like to log in again?
Total PSA (Human) ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT146-0001
75802-908EA 366.87 USD InStock
75802-908
Total PSA (Human) ELISA Kit, Enzo Life Sciences
Assays ELISAs
Highly sensitive Total PSA ELISA kit enabling detection of Total PSA in human serum, plasma, urine, and tissue culture media samples in just 2.5 hours.
  • Highly-sensitive measurement of Total PSA, detecting as little as 0.287 pM (or 0.024 ng/ml) with negligible cross reactivity with similar proteins (KLK2, ACT)
  • Compatible with various sample types (plasma, serum, urine, tissue culture media)
  • High throughput format with results in 2.5 hours for up to 40 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of Total PSA in human serum, plasma, urine and tissue culture media.
The Total PSA (human) ELISA kit is a colorimetric immunometric immunoassay kit with results in 2.5 hours.
Ordering information: Contains Microtiter plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Antibody Diluent, Conjugate, Substrate, and Stop Solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty