Would you like to log in again?
Melatonin ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT150-0001
75786-350EA 780.36 USD InStock
75786-350
Melatonin ELISA Kit, Enzo Life Sciences
Assays ELISAs
Ultra-sensitive Melatonin ELISA kit enabling detection of Melatonin in saliva, serum, plasma, and fruit homogenates in just 2 hours.
  • Sensitive measurement of Melatonin, detecting as little as 0.162 ng/ml
  • Broad dynamic range suitable for a large variety of samples
  • High throughput format with results in 2 hours for up to 40 samples in duplicate
  • Validated for a variety of sample matrices including fruit homogenates
For the quantitative determination of melatonin in any species
The Melatonin ELISA kit is a colorimetric competitive immunoassay kit with results in 2 hours.
Validated specifically for saliva, serum, plasma, and fruit samples.
Ordering information: Contains Goat anti-mouse IgG Microtiter Plate, Assay Buffer 10, Standard, Wash Buffer Concentrate, Antibody, Tracer, Conjugate, TMB Substrate, and Stop Solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty