Would you like to log in again?
EQ2 NHS ester Amine-reactive quencher dye
Supplier: Enzo Life Sciences
CHM166-0001
10810-048EA 132.56 USD InStock
10810-048
EQ2 NHS ester Amine-reactive quencher dye
EQ2 NHS ester
Amine-reactive quencher dye has a broad visible absorption but no detectable fluorescence emission in fluorescence resonance energy transfer (FRET) applications.
  • Commonly used in molecular biology FRET applications to quench fluorophores
  • NHS ester quencher dye can be conjugated to amine-labeled probes
  • Enable multiplexing of probes with the use of different reporter-quencher dye pairs
  • Compatible with most common commercially available fluorophores
EQ2 NHS ester is an amine-reactive quencher dye. It has a broad visible absorption but no detectable fluorescence emission, making it useful as an acceptor in fluorescence resonance energy transfer (FRET) applications.
It can be used to quench reporter dyes: TET, JOE, VIC, HEX, Cy3, NED, TAMRA, Cy3.5, ROX, Texas Red, Cy5, Quasar® 570, Pulsar® 650, CAL Fluor® Gold 540, CAL Fluor® Orange 560, CAL Fluor® Red 590, CAL Fluor® Red 610, CAL Fluor® Red 635, Alexa Fluor® 532, Alexa Fluor® 546, Alexa Fluor® 555, Alexa Fluor® 568, Alexa Fluor® 594, Alexa Fluor® 610, Alexa Fluor® 633, Alexa Fluor® 647, DyLight® 550, DyLight® 594, DyLight® 633, DyLight® 650, ATTO 520, ATTO 532, ATTO Rho6G, ATTO 550, ATTO 565, ATTO Rho11, ATTO 590, ATTO 594, ATTO 633, ATTO Rho14, and ATTO 647.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty