Would you like to log in again?
Lysosome Purification Kit, BioVision Inc.
Supplier: BioVision
AB234047-50TEST
75836-730EA 846.42 USD InStock
75836-730
Lysosome Purification Kit, BioVision Inc.
Assays Cellular Assays
Simple method to isolate enriched or purified lysosomes from tissue and cultured cells.
  • Simple & convenient protocol
  • The kit provides unique formulations of ready-to-use buffers and reagents to isolate lysosomes with minimal sample preparation
  • Isolation of high purity lysosomes from tissues and cultured cells
Lysosomes are membrane-bound cell organelles found in most animal cells (except red blood cells). Lysosomes are spherical vesicles containing hydrolytic enzymes capable of breaking down different kinds of biomolecules, including proteins, nucleic acids, carbohydrates, lipids, and cellular debris. They are known to contain over 50 different enzymes, which are active under acidic conditions. Thus, lysosomes act as the waste disposal system of the cells by digesting unwanted materials in the cytoplasm. BioVision’s Lysosome Isolation Kit provides a procedure for isolating enriched or purified lysosomal fraction from animal tissues and cultured cells by differential centrifugation followed by density gradient centrifugation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty