Would you like to log in again?
Live/Dead Cell Viability Assay Kit (for Mammalian Cells), BioVision
Supplier: BioVision
AB287858-96TEST
75836-742EA 823.95 USD InStock
75836-742
Live/Dead Cell Viability Assay Kit (for Mammalian Cells), BioVision
Assays Cellular Assays
Non-radioactive kit to measure cell viability, cytotoxicity & apoptosis by microscopy/FACS analysis.
  • Easy to use Protocol
  • Non-radioactive detection
  • Measure Cell Proliferation, cell viability, chemotaxis, cytotoxicity and apoptosis
  • Detect using either microscopy or FACS
Quantification of number of live and dead cells is an indispensable tool in cell biology research. BioVision’s Live/Dead Cell Viability Assay Kit, provides a two-color fluorescence method that is based on the simultaneous determination of live and dead cells using two different dyes. Live cell dye easily penetrates intact, live cells and intracellular esterase hydrolyzes the dye to produce a hydrophilic, strongly fluorescent compound that is retained in the cell cytoplasm which can be measured at Ex/Em = 485/530 nm. Dead cell dye enters damaged cell membranes and undergoes a 40-fold enhancement of fluorescence upon binding to nucleic acid, thereby producing a bright red fluorescence (Ex/Em = 495/635 nm) in dead cells. This assay kit provides an easy-to-use, non-radioactive, histological and FACS-based method for measuring cell proliferation, cell viability, chemotaxis, cytotoxicity and apoptosis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty