Would you like to log in again?
Cytomegalovirus (CMV) IgG ELISA Kit, BioVision
Supplier: BioVision
AB285253-96TEST
75837-062EA 949.95 USD InStock
75837-062
Cytomegalovirus (CMV) IgG ELISA Kit, BioVision
Assays ELISAs
ELISA kit that detect the precense of Cytomegalovirus IgG.
  • Easy, convenient and time-saving method to detect CMV IgG antibody in both primary CMV infections and in reactivated infection
  • Application- Detect the precense of Cytomegalovirus IgG in human serum or plasma
Cytomegalovirus (CMV) is a member of the herpes group of viruses. Most adults and children who catch CMV have no symptoms and are not harmed by the virus. CMV infection is of clinical significance primarily in pregnant women, newborn infants with possible congenital infection, immunosuppressed transplant patients and individuals with AIDS. CMV is so prevalent as over 60% of people catch the infection at some time in their lives. Significant increases in CMV IgG antibody by ELISA suggest recent infection or reactivation of a latent CMV infection. ELISA can detect CMV IgM antibody in both primary CMV infections (93-100%) and in reactivated infection (40%). An IgM response may be reduced or absent in immunocompromised patients with active infection. In transplant patients the CMV infection can be associated with higher morbidity and mortality.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty