Would you like to log in again?
Cotinine ELISA Kit, BioVision Inc.
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AB285286-96TEST
75837-184EA 1273.76 USD InStock
75837-184
Cotinine ELISA Kit, BioVision Inc.
Assays ELISAs
ELISA kit that measure cotinine level, a reliable marker for smoking status and cessation studies.
  • Easy, convenient and time-saving method to measure the level of cotinine in human serum and urine to determin the smoking status and smoking cessation
  • Application: Detect cotinine in serum and urine
Exposure to tobacco smoke can be detected by measuring nicotine and its metabolites. Nicotine has a short half life and is not used as a marker for tobacco smoke exposure. Cotinine due to its longer half life has been used in research as a reliable marker for smoking status and smoking cessation studies. BioVision’s Cotinine Direct ELISA Kit is designed for the detection Cotinine in serum and urine. It can also be adapted for other fluids.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty