Would you like to log in again?
Bacterial Strain JM109, Promega
Supplier: Promega
P9751
PAP9751EA 65.17 USD InStock
PAP9751
Bacterial Strain JM109, Promega
Nucleic Acid Reagents Cloning Kits and Ligation
Useful host for transformation of pGEM Vectors and production of ssDNA from M18 or phagemid vectors. JM109 is recA- and lacks the E. coli K restriction system. Can be used for blue/white screening.
  • Useful Host for Transforming Plasmids Capable of Blue/White Screening
  • Reliable strain that grows well and is transformed efficiently
  • Useful for cloning, single-stranded DNA production, and blue/white screening
  • Yield and quality of plasmid DNA improved with the endonuclease A- mutation
Bacterial Strain JM109 is a useful host for transformation of pGEM Vectors and for production of single-stranded DNA from M13 or phagemid vectors. The strain grows well and is transformed efficiently by a variety of methods. Because JM109 is recA- and lacks the E. coli K restriction system, undesirable restriction of cloned DNA and recombination with host chromosomal DNA are prevented. The endonuclease A- mutation leads to an improved yield and quality of isolated plasmid DNA. JM109 is deficient in beta-galactosidase activity due to deletions in both genomic and episomal copies of the lacZ gene. The deletion in the episomal (F' factor) copy of the lacZ gene (lacZDeltaM15) can be complemented by addition of a functional alpha-peptide encoded by a pGEM-Z or pGEM-Zf Vector. If complementation does not occur, bacterial colonies are white. To maintain the F', JM109 should be grown on minimal (M-9) media supplemented with 1mM thiamine. Genotype: endA1, recA1, gyrA96, thi, hsdR17 (r(k)-, m(k)+), relA1, supE44, Delta(lac-proAB), [F' traD36, proAB, laqI(q)ZDeltaM15].
The endonuclease A– mutation leads to an improved yield and quality of isolated plasmid DNA.
Because JM109 is recA– and lacks the E. coli K restriction system, undesirable restriction of cloned DNA and recombination with host chromosomal DNA are prevented.
Caution: Storage Conditions: Less than -65°C
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty