Would you like to log in again?
Hemoglobin Colorimetric Assay Kit, BioVision Inc.
Supplier: BioVision
AB234046-200TEST
75836-722EA 668.12 USD InStock
75836-722
Hemoglobin Colorimetric Assay Kit, BioVision Inc.
Assays Protein Assays
A simple, sensitive & easy colorimetric assay to quantitate hemoglobin in human samples.
  • Simple one-step procedure; takes only 15 minutes
  • Fast and convenient
  • Detects physiological concentrations of hemoglobins in human samples
  • HTP adaptable
Hemoglobin (Hb) is an iron containing globular metalloprotein found primarily in red blood cells. It carries oxygen from the lungs to the rest of the body as oxyhemoglobin and then returns back to the lungs as deoxyhemoglobin. In most vertebrates, hemoglobin is in tetrameric form comprising two α-globin chains and two β-globin chains (α2β2). Each chain is associated with a non-protein heme group. Aside from oxygen transport, hemoglobin can bind and transport other molecules like nitric oxide and carbon monoxide. Hemoglobin also plays an important role in maintaining the shape of the red blood cells. Alterations in the structure and binding capacity of hemoglobin are associated with many blood disorders such as sickle-cell anemia and thalassemia. BioVision’s Hemoglobin Colorimetric Assay kit provides a quick and easy method for monitoring hemoglobin levels in a wide variety of samples. In this assay, the detector selectively converts heme into a stable chemical complex that absorbs maximally at 575 nm. The intensity of the color is directly proportional to the total concentration of hemoglobin in the sample. The kit can detect as low as 0.02 g/dl hemoglobin.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty