Would you like to log in again?
Factor VIIIa Activity Assay Kit (Fluorometric), BioVision Inc.
Supplier: BioVision
AB204696-100TEST
75836-736EA 943.98 USD InStock
75836-736
Factor VIIIa Activity Assay Kit (Fluorometric), BioVision Inc.
Assays Protein Assays
Convenient assay to measure Factor VIIIa activity of plasma and serum samples or pure Factor VIIIa.
  • Convenient, and rapid
  • The assay can detect activity as low as 1 ng of FVIIIa in a variety of samples
The coagulation Factor VIII, also known as anti-hemophilic factor (AHF) is a vital blood-clotting protein. Factor VIII circulates in the bloodstream as an inactive protein, bound to a large multimeric glycoprotein called von Willebrand factor (VWF). Upon an injury to the blood vessel, VWF dissociates from FVIII and releases the active form of FVIII (FVIIIa). In the presence of calcium ions and negatively charged membrane phospholipids, activated factor VIII (FVIIIa) then binds to the activated Factor IX (FIXa) and proteolytically activates factor X (FX) to factor Xa (FXa). BioVision’s Factor VIIIa Activity Assay kit is based on the ability of FVIIIa to generate FXa. The generated FXa proteolytically cleaves a synthetic substrate and releases a fluorophore, AMC, which can be easily quantified by fluorescence microplate reader. The assay is simple, rapid and can detect activity as low as 1 ng of FVIIIa in a variety of samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty