Would you like to log in again?
Cytochrome P450 3A4 (CYP3A4) Activity Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB211076-200TEST
75836-748EA 873.61 USD InStock
75836-748
Cytochrome P450 3A4 (CYP3A4) Activity Assay Kit (Fluorometric), BioVision
Assays Protein Assays
Highly Sensitive; Includes human CYP3A4 - NADPH Reductase as positive control.
  • Simple, convenient, highly sensitive
  • Applications -Rapid assessment of native/recombinant CYP3A4 activity in lysates or microsomal fractions prepared from tissues, cells
  • Screening of drugs and novel ligands for interaction with native/recombinant CYP3A4
Cytochrome P450 3A4 (CYP3A4, EC 1.14.13.157) is a member of the cytochrome P450 monooxidase (CYP) family of microsomal xenobiotic metabolism enzymes. CYPs are membrane-bound hemeproteins responsible for Phase I biotransformation reactions, in which lipophilic drugs and other xenobiotic compounds are transformed to more hydrophilic products to facilitate excretion from the body. CYP3A4 is expressed in high levels in the liver and intestines, where it catalyzes oxidation of an extraordinarily wide variety of structurally distinct ligands. More than half of all small molecule drugs commonly used by humans are metabolized by CYP3A4. Inhibition of CYP3A4-mediated metabolism is a common cause of adverse drug/drug and drug/food interactions and toxicity. In addition, for drugs whose pharmacological activity requires metabolism from a pro-drug form, CYP3A4 inhibition can lead to decreased drug efficacy. BioVision’s CYP3A4 Activity Assay Kit enables rapid measurement of native or recombinant CYP3A4 activity in biological samples such as liver microsomes. The assay utilizes a non-fluorescent CYP3A4 substrate that is converted into a highly fluorescent metabolite detected in the visible range (Ex/Em = 535/587 nm), ensuring a high signal-to-background ratio with little interference by autofluorescence. CYP3A4 specific activity is calculated by running parallel reactions in the presence and absence of the potent inhibitor Ketoconazole and subtracting any residual activity detected with the inhibitor present. The kit contains a complete set of reagents sufficient for performing 100 sets of paired reactions (in the presence and absence of inhibitor).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty