Would you like to log in again?
Cytochrome P450 3A4 (CYP3A4) Inhibitor Screening Kit (Fluorometric), BioVision
Supplier: BioVision
AB211077-200TEST
75836-750EA 1072.52 USD InStock
75836-750
Cytochrome P450 3A4 (CYP3A4) Inhibitor Screening Kit (Fluorometric), BioVision
Assays Protein Assays
High-throughput compatible, quick screening for CYP3A inhibitors or ligands; Includes human CYP3A4.
  • Simple, highly sensitive, high-throughput compatible
  • Rapid screening of CYP3A4 inhibitors or ligands
  • Kit includes the canonical CYP3A inhibitor ketoconazole and a stable, recombinant human CYP3A4 co-expressed with NADPH Reductase
Cytochrome P450 3A4 (CYP3A4, EC 1.14.13.157) is a member of the cytochrome P450 monooxidase (CYP) family of microsomal xenobiotic metabolism enzymes. CYPs are membrane-bound hemeproteins responsible for Phase I biotransformation reactions, in which lipophilic drugs and other xenobiotic compounds are transformed to more hydrophilic products to facilitate excretion from the body. CYP3A4 is expressed in high levels in the liver and intestines, where it catalyzes oxidation of an extraordinarily wide variety of structurally distinct ligands. More than half of all small molecule drugs commonly used by humans are metabolized by CYP3A4 and inhibition of CYP3A4-mediated metabolism is a common cause of adverse drug/drug and drug/food interactions and toxicity. In addition, for drugs whose pharmacological activity requires metabolism from a pro-drug form, CYP3A4 inhibition can lead to decreased drug efficacy. BioVision’s CYP3A4 Inhibitor Screening Kit enables rapid screening of drugs and other new chemical entities (NCEs) for compound-CYP3A4 interaction in a reliable, high-throughput fluorescence-based assay. The kit provides a yeast microsomal preparation of human CYP3A4 and cytochrome P450 reductase (CPR) enzymes. The assay utilizes a non-fluorescent CYP3A4 substrate that is converted into a highly fluorescent metabolite detected in the visible range (Ex/Em = 535/587 nm), ensuring a high signal-to-background ratio with little interference by autofluorescence. The concentration of fluorogenic substrate used for screening is roughly equivalent to its Km for CYP3A4, facilitating detection of weak competitive inhibitors. The kit contains a complete set of reagents sufficient for performing 200 reactions in a 96-well plate format.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty