Would you like to log in again?
HIV-1 Protease Inhibitor Screening Kit (Fluorometric), BioVision Inc.
Supplier: BioVision
AB211106-100TEST
75836-758EA 980.33 USD InStock
75836-758
HIV-1 Protease Inhibitor Screening Kit (Fluorometric), BioVision Inc.
Assays Protein Assays
Market's first and only kit for screening HIV-1 Protease Inhibitors
  • Simple and High throughput adaptable
  • Detect using a fluorometer or fluorescence microplate reader.
  • The simple and high-throughput adaptable assay kit can be used to screen/study/characterize potential inhibitors of HIV-1 Protease with a fluorometer or fluorescence microplate reader.
Human Immunodeficiency Virus (HIV) is the cause of the Acquired Immunodeficiency Syndrome (AIDS). HIV-1 protease is a retroviral aspartyl protease (retropepsin) that is essential for the life-cycle of the virus as it cleaves newly synthesized polyproteins at the appropriate places to create the mature protein components of an infectious HIV virion. Without effective HIV-1 protease, HIV virions remain non-infectious. BioVision’s HIV-1 Protease Inhibitor Screening kit utilizes the ability of active HIV-1 protease to cleave a synthetic peptide substrate resulting in the release of free fluorophore, which can be easily quantified using a fluorometer or fluorescence microplate reader. In the presence of a HIV-1 Protease inhibitor, the cleavage of the substrate is reduced/abolished resulting in decrease or total loss of the fluorescence. This simple and high-throughput adaptable assay kit can be used to screen/study/characterize potential inhibitors of HIV-1 Protease.
Simple and HTP compatible.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty