Would you like to log in again?
Cytochrome P450 1A2 (CYP1A2) Activity Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB211074-100TEST
75837-068EA 643.53 USD InStock
75837-068
Cytochrome P450 1A2 (CYP1A2) Activity Assay Kit (Fluorometric), BioVision
Assays Protein Assays
Highly Sensitive; Includes human CYP1A2 - NADPH Reductase as positive control.
  • Simple, convenient, highly sensitive
  • Fluorescent assay enables easy determination of CYP1A2 activity in a variety of biological samples
  • The substrate shows low background and a high signal-to-noise ratio
  • Kit includes CYP1A2 inhibitor (α-naphthoflavone) and a stable, recombinant human CYP1A2 co-expressed with NADPH Reductase as a positive control
Cytochrome P450 1A2 (CYP1A2, EC 1.14.14.1) is a member of the cytochrome P450 monooxidase (CYP) family of microsomal xenobiotic metabolism enzymes. CYPs are membrane-bound hemeproteins responsible for Phase I biotransformation reactions, in which lipophilic drugs and other xenobiotic compounds are converted to more hydrophilic products to facilitate excretion from the body. CYP1A2 is primarily expressed in liver, intestinal and olfactory mucosal tissue and catalyzes oxidation of polyaromatic and heterocyclic molecules such as aromatic amines. CYP1A2 is responsible for metabolism of approximately 10% of all small molecule drugs commonly used by humans. Polymorphisms in the human CYP1A2 gene have been implicated in clinical drug/drug interactions involving widely-used drugs, including caffeine, theophylline and the antipsychotic clozapine. Isoforms of the CYP1A subfamily are also involved in metabolic activation of environmental pro-carcinogens present in cigarette smoke and combustion exhaust fumes. BioVision’s CYP1A2 Activity Assay Kit enables rapid measurement of native or recombinant CYP1A2 activity in biological samples such as liver microsomes. The assay utilizes a non-fluorescent CYP1A2 substrate that is converted into a highly fluorescent metabolite detected in the visible range (Ex/Em = 406/468 nm), ensuring a high signal-to-background ratio with little interference by autofluorescence. A selective CYP1A2 inhibitor is provided for determination of CYP1A2 activity in heterogeneous biological samples, where other CYP isozymes may contribute to substrate metabolism. The inhibitor displays greater than 20-fold selectivity for CYP1A2 over other CYPs, ensuring targeted inhibition. CYP1A2 specific activity is calculated by running parallel reactions in the presence and absence of the selective inhibitor and subtracting any residual activity detected with the inhibitor present. The kit contains a complete set of reagents sufficient for performing 50 sets of paired reactions (in the presence and absence of inhibitor).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty