Would you like to log in again?
Cytochrome P450 1A2 (CYP1A2) Inhibitor Screening Kit (Fluorometric), BioVision
Supplier: BioVision
AB211075-100TEST
75837-070EA 720.12 USD InStock
75837-070
Cytochrome P450 1A2 (CYP1A2) Inhibitor Screening Kit (Fluorometric), BioVision
Assays Protein Assays
High-throughput compatible, quick screening for CYP1A2 inhibitors or ligands; Includes human CYP1A2
  • Simple, highly sensitive, high-throughput compatible
  • Rapid screening of CYP1A2 inhibitors or ligands
  • Kit includes the CYP1A2 inhibitor α-naphthoflavone and a stable, recombinant human CYP1A2 co-expressed with NADPH Reductase
Cytochrome P450 1A2 (CYP1A2, EC 1.14.14.1) is a member of the cytochrome P450 monooxidase (CYP) family of microsomal xenobiotic metabolism enzymes. CYPs are membrane-bound hemeproteins responsible for Phase I biotransformation reactions, in which lipophilic drugs and other xenobiotic compounds are converted to more hydrophilic products to facilitate excretion from the body. CYP1A2 is primarily expressed in liver, intestinal and olfactory mucosal tissue and catalyzes oxidation of planar polyaromatic and heterocyclic molecules such as aromatic amines. CYP1A2 is responsible for metabolism of approximately 10% of all small molecule drugs commonly used by humans. Polymorphisms in the human CYP1A2 gene have been implicated in clinical drug/drug interactions involving widely-used drugs, including methylxanthines (caffeine and theophylline), ciprofloxacin and a number of antidepressants and antipsychotics. Isoforms of the CYP1A subfamily are also involved in metabolic activation of environmental pro-carcinogens in cigarette smoke and combustion exhaust fumes. BioVision’s CYP1A2 Inhibitor Screening Kit enables rapid screening of drugs and other new chemical entities (NCEs) for compound-CYP1A2 interaction in a reliable, high-throughput fluorescence-based assay. The kit provides a yeast microsomal preparation of human CYP1A2 and human cytochrome P450 reductase (CPR) enzymes. The assay utilizes a non-fluorescent CYP1A2 substrate that is converted into a highly fluorescent metabolite detected in the visible range (Ex/Em = 406/468 nm), ensuring a high signal-to-background ratio with little interference by autofluorescence. The kit contains a complete set of reagents sufficient for performing 100 reactions in a 96-well plate format.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty