Would you like to log in again?
EZLabel™ Antibody FITC Labeling Kit, BioVision
Supplier: BioVision
AB288100-5TEST
75836-760EA 324.11 USD InStock
75836-760
EZLabel™ Antibody FITC Labeling Kit, BioVision
Protein Linking and Labeling Systems
Easy to use Kit to label upto 5mg of AB wz FITC for multiple application such as ELISA,western blot.
  • All necessary reagents included.
  • Fast and efficient labeling of antibodies with FITC
  • Easy to use Kit to label upto 5 mg of Antibody with FITC for multiple applications such as ELISA, western blot, Immunohistochemistry, Immunofluorescence and FACS analysis
BioVision’s EZLabel Antibody-FITC Labeling Kit provides an easy way to label antibodies with Fluorescein Isothiocyanate (FITC) in a user-friendly spin column format. FITC is an ideal dye for fluorescent labeling of antibodies among the most commonly used fluorescent dyes for labeling antibodies. Each spin column provided in the kit can be used to label up to 1 mg of the target antibody. The kit provides all of the reagents necessary to perform five labeling reactions using up to 1 mg of antibody per reaction. FITC-labeled antibody has an excitation and emission wavelengths at 494 nm and 520 nm respectively, and can be directly used for downstream applications including ELISA, western blot, Immunohistochemistry, Immunofluorescence, and FACS analysis, etc.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty