Would you like to log in again?
EZLabel™ Antibody Biotin Labeling Kit, BioVision
Supplier: BioVision
AB288097-5TEST
75837-046EA 329.52 USD InStock
75837-046
EZLabel™ Antibody Biotin Labeling Kit, BioVision
Protein Linking and Labeling Systems
Easy to use Kit to label upto 5mg of AB wz Biotin for multiple applications such as ELISA, Wblot.
  • All necessary reagents included.
  • Fast and efficient labeling of antibodies with Biotin
  • Easy to use Kit to label upto 5 mg of Antibody with Biotin for multiple applications such as ELISA, western blot, Immunohistochemistry, Immunofluorescence and FACS analysis
BioVision’s EZLabel™ Antibody-Biotin Labeling Kit provides an easy way to label antibodies with Biotin in a user-friendly spin column format. Due to its high affinity for tetrameric avidin proteins, biotin is ideal for labeling antibodies. Each pair of spin columns provided in the kit can be used to purify up to 1 mg of the labeled target antibody. The kit provides all of the reagents necessary to perform five labeling reactions using up to 1 mg of antibody per reaction. The biotin-labeled antibody can be directly used for multiple downstream applications including ELISA, western blot, Immunohistochemistry, Immunoassays, etc.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty