Would you like to log in again?
EZLabel™ Antibody Cy3 Labeling Kit, BioVision
Supplier: BioVision
AB288108-5TEST
75837-050EA 306.44 USD InStock
75837-050
EZLabel™ Antibody Cy3 Labeling Kit, BioVision
Protein Linking and Labeling Systems
Easy to use Kit to label upto 5 mg of Antibody wz Cy3 for multiple application such as ELISA, wblot.
  • All necessary reagents included.
  • Fast and efficient labeling of antibodies with Cy3
  • Easy to use Kit to label upto 5 mg of Antibody with Cy3 for multiple applications such as ELISA, western blot, Immunohistochemistry, Immunofluorescence and FACS analysis
BioVision’s EZLabel™ Antibody-Cy3 Labeling Kit provides an easy way to label antibodies with Cy3 in a user-friendly spin column format. Cy3 is a better alternative to Alexa Fluor 555, Rhodamine Red-X, and TAMRA due to its high extinction coefficient and far red emission. Each pair of spin columns provided in the kit can be used to purify up to 1 mg of the labeled target antibody. The kit provides all of the reagents necessary to perform five labeling reactions using up to 1 mg of antibody per reaction. Cy3-labeled antibody has excitation and emission wavelengths at 550 nm and 570 nm respectively, and can be directly used for multiple downstream applications including ELISA, western blot, Immunohistochemistry, Immunofluorescence, FACS, etc.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty