Would you like to log in again?
BBL™ Nutrient Agar, BD
Supplier: BD Life Sciences
BBL™
220971 297801 297801
90008-680PK 62.3 USD InStock
90008-680 CA90008-680 90001-172
BBL™ Nutrient Agar, BD
Media for Microbiology Prepared Media for Microbiology
Nutrient Agar is a general-purpose medium for the cultivation of a wide variety of bacterial organisms.
  • Peptones provide organic nitrogen and amino acids
  • Beef extract provides carbohydrates, vitamins, nitrogen, and salts
  • Agar is the solidifying agent
This relatively simple formulation provides the nutrients necessary for the replication of a large number of microorganisms which are not excessively fastidious.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty