Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fluid K, BD Diagnostics

Supplier: BD Diagnostics
215178
90008-993PK 569.48 USD InStock
90008-993
Fluid K, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Diluting Fluid K is recommended for sterility testing of pharmaceuticals in accordance with USP 2008.

  • Prepared appearance: Light yellow-colored clear solution
  • pH 6.7-7.1
  • Contains peptic digest, beef extract, and polysorbate 80

Diluting Fluid K is recommended as rinsing fluid for membrane filter method used in validation tests for bacteriostasis and fungistasis activity of pharmaceutical articles before carrying out sterility test procedures as per USP.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty