Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Native Bovine Trypsin (from Pancreas)
Supplier: MilliporeSigma
6502-25MU
80057-678EA 611.85 USD InStock
80057-678
Native Bovine Trypsin (from Pancreas)
Enzymes
Native trypsin from bovine pancreas, serine protease that specifically hydrolyzes amides, peptides and proteins at the C-terminal side of arginine and lysine residues
Optimal pH of 8.5 to 8.8.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty