Would you like to log in again?
BBL™ Sabouraud Dextrose Agar Deeps (Pour Tube), BD
Supplier: BD Life Sciences
296182 296182
90004-660PK 666.66 USD InStock
CA-90004-660 90004-660
BBL™ Sabouraud Dextrose Agar Deeps (Pour Tube), BD
Media for Microbiology Prepared Media for Microbiology
Sabouraud Dextrose Agar is a nonselective medium for the cultivation and maintenance of pathogenic and non-pathogenic fungi, particularly dermatophytes.
  • Peptones provide a source of nitrogenous growth factors
  • Dextrose provides an energy source
  • Agar is the solidifying agent
Selectivity is achieved by the addition of chloramphenicol.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty