Would you like to log in again?
SP Wilmad-LabGlass Adsorption Columns, SP Industries
Supplier: SP Industries
LG-5931-102
14206-726EA 856.61 USD InStock
14206-726
SP Wilmad-LabGlass Adsorption Columns, SP Industries
Distillation Components Distillation Columns
These borosilicate glass indicator adsorption columns are used to determine saturates, nonaromatic olefins, and aromatics in petroleum fractions that distill below 315˚C, as specified in ASTM Method D-1319
The standard flourescent indicator column is drawn to a fine point for retaining silica gel. The precision engineered flourescent indicator column has a removable capillary tip joined to the analyzer section by a 12/2 spherical joint. Columns have a 28/12 socket joint at the top.
Certifications: Reference No. ASTM D 1319 Standard Method for Hydrocarbon Types in Liquid Petroleum Products by Fluorescent Indicator Adsorption.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty