Would you like to log in again?
BBL™ Sabouraud Liquid Broth, Modified (Antibiotic Assay Medium #13), BD
Supplier: BD Life Sciences
221014 221014
90001-188PK 56.72 USD InStock
CA90001-188 90001-188
BBL™ Sabouraud Liquid Broth, Modified (Antibiotic Assay Medium #13), BD
Media for Microbiology Prepared Media for Microbiology
Sabouraud Liquid Broth, Modified is a medium for the cultivation of fungi.
  • Casein and meat peptones provide source of nitrogen and carbon
  • Dextrose is a source of energy
  • Made according to USP specifications
The concentration of antibiotic is determined by comparing amounts of growth obtained with that given by the reference standard solutions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty