Would you like to log in again?
ToxOut™ Rapid Endotoxin Removal Regeneration Buffer, BioVision
Supplier: BioVision
AB269281-50ML
75838-182EA 360.04 USD InStock
75838-182
ToxOut™ Rapid Endotoxin Removal Regeneration Buffer, BioVision
Buffers Biological Buffers
50 ml of Regeneration buffer for quickly regenerating BioVision‘s ToxOut Rapid Endotoxin Removal Agarose.
  • Endotoxin free
  • Can quickly and effectively eliminate endotoxins
  • 50 ml of Regeneration buffer for ToxOut Rapid Endotoxin Removal Agarose
Endotoxin is the lipopolysaccharide (LPS) complex located in the outer membrane of gram-negative bacteria. During experimental procedures, large amount of endotoxins are shed and can easily contaminate labware, buffers and downstream products. Presence of endotoxin in any of these has a detrimental effect on culturing of cells and can lead to generation of irreproducible or negative experimental data. Therefore, it is critical to remove endotoxin from samples and products. ToxOutTM Rapid Endotoxin Removal Regeneration Buffer is the endotoxin-free regeneration buffer containing 1% sodium deoxycholate. It is specially designed for quickly regenerating BioVision‘s ToxOutTM Rapid Endotoxin Removal Agarose, which can quickly and effectively eliminate endotoxins to < 0.05 EU/mL in solutions containing proteins or pharmacologically important components.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty