Would you like to log in again?
ToxOut™ Endotoxin Removal Equilibration Buffer, BioVision
Supplier: BioVision
AB278802-50ML
75838-184EA 267.73 USD InStock
75838-184
ToxOut™ Endotoxin Removal Equilibration Buffer, BioVision
Buffers Biological Buffers
50 ml of Regeneration buffer for ToxOut Endotoxin Removal Agarose to neutral pH for better detox efficiency and protein recovery.
  • Rapid Endotoxin Removal Agarose
  • High capacity endotoxin removal Agarose, Better Detox
  • Efficiency and protein recovery
Endotoxin is the lipopolysaccharide (LPS) complex located in the outer membrane of gram-negative bacteria. During experimental procedures, large amount of endotoxins are shed and can easily contaminate labware, buffers and downstream products. Presence of endotoxin in any of these has a detrimental effect on culturing of cells and can lead to generation of irreproducible or negative experimental data. Therefore, it is critical to remove endotoxin from samples and products. ToxOutTM Endotoxin Removal Equilibration Buffer is designed to equilibrate BioVision‘s ToxOutTM Rapid Endotoxin Removal Agarose and ToxOutTM High Capacity Endotoxin Removal Agarose to neutral pH for better detox efficiency and protein recovery. Both ToxOutTM Rapid Endotoxin Removal Agarose and ToxOutTM High Capacity Endotoxin Removal Agarose can effectively eliminate endotoxins to ≤ 0.05 EU/mL in solutions containing proteins or pharmacologically important components.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty