Would you like to log in again?
Vitamin D, 25-OH Total RIA, IBL America
Supplier: IBL America
KIR1971
102968-328EA 1023.51 USD InStock
102968-328
Vitamin D, 25-OH Total RIA, IBL America
Assays Immunological Assays
Radioimmunoassay (RIA) for the determination of 25-hydroxyvitamin D3 and D2 (25-OH-D3 and 25-OH-D2) in serum.
  • For research use only, not for use in diagnostic procedures
  • A competitive RIA assay
  • Novel pre-treatment step eliminates the use of organic solvent precipitation and centrifugation
The 25OH Vitamin D Total RIA is a competitive RIA assay, based on patented monoclonal antibodies, with a novel pre-treatment step performed inside the RIA coated tubes.
The novel pre-treatment step of the 25OH Vitamin D Total RIA takes place directly in the coated tubes. It uses a unique displacement reagent that displaces the 25OH Vit D from the binding proteins. The advantage of the technique is that it works at neutral pH so it can be used in the presence of the antibodies, which is not the case for the other known methods, most of them using acidic or alkaline pH.
Furthermore, this novel pre-treatment step, which eliminates the use of organic solvent precipitation and centrifugation, highly facilitates the automation on e.g. RIAMAT, Tecan or BioTek pipetting systems. The assay uses small sample volume (25 µL) which makes it particularly suitable for pediatric samples. The incubation steps are performed at room temperature without need for dedicated equipment.
The assay is calibrated to the ID-LC-MS/MS reference method developed by Prof. Dr. Linda THIENPONT from the Ghent University in Belgium, as encompassed in the Vitamin D Standardization Program initiative. It shows a high correlation with other commercially available LC-MS/MS and RIA assays.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty