Would you like to log in again?
Vitamin D3, 1-25-OH RIA, IBL America
Supplier: IBL America
KIR1929
103123-436EA 1110.15 USD InStock
103123-436
Vitamin D3, 1-25-OH RIA, IBL America
Assays Immunological Assays
Radioimmunoassay (RIA) for the determination of human 1,25(OH)2-Vitamin D (1,25(OH)2-Vit.D) in serum and plasma
  • For research use only, not for use in diagnostic procedures
  • Determineof human 1,25(OH)2-Vitamin D (1,25(OH)2-Vit.D) in serum and plasma
Radioimmunoassay (RIA) for the determination of human 1,25(OH)2-Vitamin D (1,25(OH)2-Vit.D) in serum and plasma. For research use only, not for use in diagnostic procedures.
For research use only, not for use in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty