Would you like to log in again?
Vitamin D3, 25-OH RIA, IBL America
Supplier: IBL America
KIP1961
102968-326EA 934.15 USD InStock
102968-326
Vitamin D3, 25-OH RIA, IBL America
Assays Immunological Assays
Radioimmunoassay (RIA) for the determination of 25OH-Vit.D3 in human serum and heparin plasma.
  • For research use only, not for use in diagnostic procedures
  • Competitive RIA assay
  • Calibrated to the ID-LC-MS/MS reference method
The 25OH Vitamin D3 RIA is a competitive RIA assay, specific for the D3 form of 25OH Vitamin D.
The assay is calibrated to the ID-LC-MS/MS reference method developed by Prof. Dr. Linda THIENPONT from the Ghent University in Belgium, as encompassed in the Vitamin D Standardization Program initiative. It shows a high correlation with other commercially available LC-MS/MS and RIA assays.
At first calibrators, controls and samples (serum or plasma) are extracted with acetonitrile. A fixed amount of 125I labelled 25OH Vitamin D3 competes with the 25OH Vitamin D3 from either extracted samples, controls or calibrators for a fixed amount of specific antibody sites immobilized to the lower and inner surface of plastic tubes.
After 2 hours incubation at room temperature, an aspiration step stops the competition reaction. The tubes are then washed with 3 ml washing solution and counted in a gamma counter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty