Would you like to log in again?
Vitamin D (25OH) Total ELISA, IBL America
Supplier: IBL America
KAP1971
102465-526EA 662.03 USD InStock
102465-526
Vitamin D (25OH) Total ELISA, IBL America
Assays ELISAs
Immunoenzymetric assay (ELISA) for the quantitative determination of 25-hydroxyvitamin D2 and D3 (25OH-D2 and 25OH-D3) in serum. 510(k) cleared.
  • Competitive ELISA assay
  • Works at neutral pH so it can be used in the presence of antibodies
  • Very competitive sensitivity, precision and performance characteristics
The 25OH Vitamin D Total ELISA is a competitive ELISA assay, based on patented monoclonal antibodies, with a novel pre-treatment step performed inside the ELISA microtiter plate.
The novel pre-treatment step of the 25OH Vitamin D Total ELISA takes place directly in the microtiter plate. It uses a displacement reagent that displaces the 25OH Vit D from the binding proteins. The advantage of the technique is that it works at neutral pH so it can be used in the presence of the antibodies, which is not the case for the other known methods, most of them using acidic or alkaline pH.
The assay shows very competitive sensitivity, precision and performance characteristics to all other immunoassays in the market but uses a novel and extremely easy pre-treatment step performed directly in the microtiter plate. The assay is calibrated to the ID-LC-MS/MS reference method developed by Prof. Dr. Linda THIENPONT from the Ghent University in Belgium, as encompassed in the Vitamin D Standardization Program initiative. It shows a high correlation with other commercially available CLIA, LC-MS/MS and ELISA assays. The 25OH Vitamin D Total ELISA was recently awarded the Certificate of Proficiency by the Vitamin D External Quality Assessment Scheme (DEQAS) Advisory Panel. This designation certifies that the assay meets the performance target set by DEQAS for 25OH Vitamin D.
The novel approach of the 25OH Vitamin D Total ELISA makes it extremely easy to program on any open ELISA platform without the need for a manual sample pre-treatment outside the ELISA automate. The assay does not require a manual transfer from extracted sample in a tube to the well in the microtiter plate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty