Would you like to log in again?
3M™ 4510 Series Disposable Protective Coveralls, 3M Company
Supplier: 3M
7000088922 7000088927 7000088926 7000088923 7100112827 7000088924
89429-122CS 486.5 USD InStock
89429-122 89429-120 89429-126 89429-124 89429-128 89429-118
3M™ 4510 Series Disposable Protective Coveralls, 3M Company
Disposable Garments
3M™ Protective Coverall 4510 is disposable safety work wear made of a high quality microporous laminated material. Suitable for various applications, it helps provide basic barrier protection against light liquid splashes and hazardous dusts.
  • Elastic waist, ankles, and wrists provide convenience and freedom of movement
  • Convenient two-way zipper with storm flap helps provide additional protection
  • Anti-static coating on both sides reduces static build-up and prevents the risk of static discharge during use
  • Garments do not contain components made from natural rubber latex to prevent reactions from users with latex sensitivities or restrictions
  • Garments do not contain components made from silicone to prevent reactions from users with silicone sensitivities or restrictions
The 4510 Series Disposable Protective Coveralls feature elastic waist, ankles, and wrists for convenience and freedom of movement
These key features make this protective coverall a great disposable PPE option. The 3M™ Protective Coverall 4510 helps protect against light liquid splashes and hazardous dusts. Typical applications for this safety work wear include asbestos abatement and inspection, exposure to radioactive particles, lead abatement, mold remediation/mold spores, fertilizer application (dry/granular), pesticide application (liquid), general industrial clean-up and processing, grease (general), light-duty building cleaning, machine or vehicle maintenance, coal dust, fiberglass/cellulose insulation installation, food processing, general powder handling, non-hazardous aerosols (sprays), non-hazardous liquids, paint/coating spraying, paint/coating spraying (with isocyanate paints), cleanroom applications (ISO Class 4–9), pharmaceutical powder handling, metal particulate (chromium beryllium, etc.), and metal polishing.
A two-way zipper, with storm flap for convenience, provides additional protection. Low-linting properties and anti-static treatment are featured on both sides. The standard collar provides convenience when using auxiliary PPE.
Certifications: Certified to European Standards to help protect against certain industrial hazards.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty