Would you like to log in again?
Streptavidin coated 96-well Plate, BioVision Inc
Supplier: BioVision
AB286859-5UNITS
75837-814EA 581.84 USD InStock
75837-814
Streptavidin coated 96-well Plate, BioVision Inc
Microplates Cell Culture Plates
Streptavidin-coated plates for detecting biotinylated proteins, Oligos or IgG.
  • These plates are useful for detecting Biotinylated proteins, oligonucleotides or IgG as low as 1 ng/ml.
  • These pre-blocked plates are ready-to-use and are compatible with fluorescent, colorimetric or chemiluminiscent detection.
  • Plates with Streptavidin coating for biotin binding
BioVision’s High Sensitivity Streptavidin plates (clear, 12x8-Well Strips), are ready-to-use biotin-binding microplates. Proprietary coating of the plates with highly pure streptavidin allows the user to detect biotin with high sensitivity and low background. The microplates are pre-blocked with BioVision’s unique blocking buffer. The plates allow detection of as low as 1 ng/ml biotinylated rabbit IgG. The plates are compatible with fluorescent, colorimetric or chemiluminescent detection, using biotinylated ligands such as biotin-labeled oligonucleotides, peptides /proteins or IgG etc.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty