Would you like to log in again?
VWR®, Air Duster Spray
Supplier: VWR
90028-11
75780-350CS 257.56 USD InStock
75780-350
VWR®, Air Duster Spray
Sprays
Clean equipment you wouldn’t dare touch.
  • Filtered for 100% purity
  • Leaves no residue
  • Moisture free
  • Odorless
  • Non-flammable/Non-conductive
  • Very low order of toxicity
Air Duster Spray is a portable, noncorrosive, nonflammable, dry and odorless gas. The “air compressor in a can” provides up to 3000 jets of cleaning power. Pressurized liquid non-flammable gas.
Duster products specifically for the removal of dust from photographic negatives, semiconductor chips, and specimens under electronic microscopes of for use on energized electrical equipment.
Certifications: DOT-1,1,1,2 Tetrafluoroethane, Non-flammable gas UN1078 DOT-E-10232, CAS No. 811-97-2. DOT-E-10232 NRC 240/300 M1119.
Caution: Facilitates compliance with ENVIRONMENTAL and OSHA standards and contains no Chlorofluorocarbons or Hydrofluorocarbons. Contents under very high pressure.
Pressure at 70°F (21°C) equals 71.2 PSIG, 100°F (38°C) equals 124.3 PSIG, and 130°F (55°C) equals 198.9 PSIG. Do not puncture or incinerate container. Do not store above 120°F (49°C).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty