Would you like to log in again?
8021/502.2 Surrogate Mix #1 (3 Components), Restek
Supplier: Restek
30463
75835-570EA 43.65 USD InStock
75835-570
8021/502.2 Surrogate Mix #1 (3 Components), Restek
Standards Environmental Analysis Standards
The mix consists of 1-bromo-2-chloroethane, 1-chloro-3-fluorobenzene and fluorobenzene.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty