Would you like to log in again?
Method 525.2 Surrogate Standard Mix (4 Components), Restek
Supplier: Restek
31826
75834-792EA 40.74 USD InStock
75834-792
Method 525.2 Surrogate Standard Mix (4 Components), Restek
Standards Environmental Analysis Standards
This mix contains 2-nitro-m-xylene, perylene-d12, pyrene-d10 and triphenylphosphate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty