Would you like to log in again?
8260A Surrogate Mix (4 components), Restek
Supplier: Restek
30240
75835-552EA 44.62 USD InStock
75835-552
8260A Surrogate Mix (4 components), Restek
Standards Environmental Analysis Standards
The surrogate mix consists of four components 1-bromo-4-fluorobenzene (BFB), dibromofluoromethane, 1,2-dichloroethane-d4 and toluene-d8.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty