Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thio-NAD

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Synonyms: Thionicotinamide Adenine dinucleotide oxidized formThionicotinamide-DPN(2R,3R,4S,5R)-5-[[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxymethyl]-2-(3-carbamothioylpyridin-1-ium-1-yl)-4-hydroxyoxolan-3-olate

AB285426-250MG AB285426-1G AB285426-50MG
75836-950EA 298.77 USD InStock
75836-950 75836-952 75836-954
Thio-NAD
Thio-NAD
An analog of NAD. Use of Thio-NAD instead of NAD (Cat. No. 2734) as a substrate for NAD(+)-consuming enzymes is more advantageous since the reduced form of Thio-NAD exhibits a substantial increase in absorbance at 405 nM, the most commonly available wavelength on microplate readers.


Formula: C₂₁H₂₇N₇O₁₃P₂S
MW: 679.491 g/mol
MDL Number: MFCD00079609
CAS Number: 4090-29-3
UN: 3316
ADR: ,
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty