Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride 0.9% in aqueous solution, sterile-filtered

Supplier: BioVision
Synonyms: SaltHaliteSea saltRock saltSaline

AB285421-500ML AB285421-100ML AB285421-1000ML
75837-684EA 293.33 USD InStock
75837-684 75837-682 75837-680
Sodium chloride 0.9% in aqueous solution, sterile-filtered
Sodium chloride
A sterile-filtered solution of 0.9% Sodium chloride in de-ionized water.


Formula: NaCl
MW: 58.44 g/mol
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty