Would you like to log in again?
Method 525.2 Nitrogen/Phosphorus Pesticide Mix #2 (6 Components), Restek
Supplier: Restek
32423
75834-970EA 85.36 USD InStock
75834-970
Method 525.2 Nitrogen/Phosphorus Pesticide Mix #2 (6 Components), Restek
Standards Organic Standards
Contains six components, those are carboxin, diazinon, disulfoton, fenamiphos, merphos and terbufos.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty