Would you like to log in again?
508.1 Calibration Mix #3 (8 Components), Restek
Supplier: Restek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
32096
75834-226EA 55.29 USD InStock
75834-226
508.1 Calibration Mix #3 (8 Components), Restek
Standards Organic Standards
Consists of eight components, those are alachlor, atrazine, chlorothalonil, cyanazine , hexachlorocyclopentadiene, metolachlor, metribuzin and simazine.
8 components
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty