Would you like to log in again?
508.1 Pesticide Kit, Restek
Supplier: Restek
32097
75834-228KT 330.77 USD InStock
75834-228
508.1 Pesticide Kit, Restek
Standards Organic Standards
Kit contains
  • 508 Performance Check Mix
  • 508.1 Internal Standard Mix
  • 508.1 Surrogate Mix
  • 508.1 GC Degradation Check Mix
  • 508.1 Calibration Mix #1
  • 508.1 Calibration Mix #2
  • 508.1 Calibration Mix #3
Ordering information: Quantity discounts not available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty