Would you like to log in again?
Chlorinated Acids by LC, Mix A (8 Components), Restek
Supplier: Restek
32431
75834-986EA 104.76 USD InStock
75834-986
Chlorinated Acids by LC, Mix A (8 Components), Restek
Standards Organic Standards
Consists of eight components, those are acifluorfen, bentazon, chloramben, 2,4-d, dicamba, dichlorprop, picloram and 2,4,5-tp.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty